missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-5-(methylcarbamoyl)benzeneboronic acid, 98%, Thermo Scientific™

Product Code. 15440426
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15440426

Brand: Thermo Scientific Alfa Aesar H53149.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is a useful intermediate in pharmaceutical intermediate. Boronic Acids are important chemical building blocks employed in cross coupling reactions. The ability of boronic acids to reversibly bind diol functional groups has been utilized for fluorescent detection of saccharides. The compound Bortezomib utilizes the boronic acid for its proteasome inhibitory effect.

Notes
Store in cool dry place. Ensure proper ventilation. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 871332-63-7
Molecular Formula C8H9BFNO3
Molecular Weight (g/mol) 196.972
MDL Number MFCD07363793
InChI Key CDJLVNMKXZMCHU-UHFFFAOYSA-N
Synonym 3-fluoro-5-methylcarbamoyl phenylboronic acid, 3-fluoro-5-methylcarbamoyl phenyl boronic acid, 3-fluoro-5-methylcarbamoyl benzeneboronic acid, n-methyl 3-borono-5-fluorobenzamide, 3-fluoro-5-methylaminocarbonyl phenylboronic acid, acmc-209qgt, 3-fluoro-5-methylcarbamoyl benzene boronic acid, 3-fluoro-5-methylcarbamoyl-phenyl boronic acid, 3-fluoro-5-methyl carbamoyl phenylboronic acid
PubChem CID 44886927
IUPAC Name [3-fluoro-5-(methylcarbamoyl)phenyl]boronic acid
SMILES B(C1=CC(=CC(=C1)F)C(=O)NC)(O)O

Specifications

Quantity 1 g
Formula Weight 196.97
Percent Purity 98%
Chemical Name or Material 3-Fluoro-5-(methylcarbamoyl)benzeneboronic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.