Learn More
3-Fluoro-4-methylbenzyl bromide, 97%, Thermo Scientific™
Product Code. 11451250
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
11451250 5 g 5g
11441250 1 g 1g
2 options
Product Code. 11451250
Brand: Thermo Scientific Alfa Aesar H26349.06
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 145075-44-1
Molecular Formula C8H8BrF
Molecular Weight (g/mol) 203.05
MDL Number MFCD01631534
InChI Key YOACSNDETIDTAY-UHFFFAOYSA-N
Synonym 3-fluoro-4-methylbenzyl bromide, 4-bromomethyl-2-fluoro-1-methylbenzene, benzene, 4-bromomethyl-2-fluoro-1-methyl, acmc-1c02p, 3-fluoro-4-methyl-benzyl bromide, 3-fluoro-4-methylbenzylbromide, benzene,4-bromomethyl-2-fluoro-1-methyl, 4-bromomethyl-2-fluoranyl-1-methyl-benzene
PubChem CID 2774603
IUPAC Name 4-(bromomethyl)-2-fluoro-1-methylbenzene
SMILES CC1=C(F)C=C(CBr)C=C1
Specifications
Boiling Point 77°C (3 mmHg)
Refractive Index 1.546
Quantity 5 g
UN Number UN3265
Formula Weight 203.06
Percent Purity 97%
Chemical Name or Material 3-Fluoro-4-methylbenzyl bromide