missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-4-methylbenzoic acid, 97%, Thermo Scientific™

Catalog Number 11469853
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
350-28-7
C8H7FO2
154.14
MFCD00002490
XUQCONCMPCVUDM-UHFFFAOYSA-N
3-fluoro-p-toluic acid, 3-fluoro-4-methylbenzoicacid, 3-fluoro-4-methyl-benzoic acid, benzoic acid, 3-fluoro-4-methyl, pubchem1330, acmc-209ich, 4-carboxy-2-fluorotoluene, 3-fluoro4-methylbenzoic acid, ksc223k8t, rarechem al bo 0761
67687
3-fluoro-4-methylbenzoic acid
CC1=CC=C(C=C1F)C(O)=O
This item is not returnable. View return policy
350-28-7
C8H7FO2
154.14
MFCD00002490
XUQCONCMPCVUDM-UHFFFAOYSA-N
3-fluoro-p-toluic acid, 3-fluoro-4-methylbenzoicacid, 3-fluoro-4-methyl-benzoic acid, benzoic acid, 3-fluoro-4-methyl, pubchem1330, acmc-209ich, 4-carboxy-2-fluorotoluene, 3-fluoro4-methylbenzoic acid, ksc223k8t, rarechem al bo 0761
67687
3-fluoro-4-methylbenzoic acid
CC1=CC=C(C=C1F)C(O)=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 169°C to 171°C
Quantity 1 g
Beilstein 2574141
Formula Weight 154.14
Percent Purity 97%
Chemical Name or Material 3-Fluoro-4-methylbenzoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'