missing translation for 'onlineSavingsMsg'
Learn More

3-Ethoxy-4-hydroxybenzaldehyde, 99.9%, MP Biomedicals™

Product Code. 11495401 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11495401 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11495401 Supplier MP Biomedicals Supplier No. 0215795980

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 121-32-4
Color White
Assay Percent Range 99.9%
Molecular Formula C9H10O3
MDL Number MFCD00006944
Quantity 100 g
Synonym ethyl vanillin, ethylvanillin, bourbonal, ethylprotal, ethavan, ethovan, vanirom, vanilal, quantrovanil, vanillal
InChI Key CBOQJANXLMLOSS-UHFFFAOYSA-N
SMILES CCOC1=CC(C=O)=CC=C1O
IUPAC Name 3-ethoxy-4-hydroxybenzaldehyde
Molecular Weight (g/mol) 166.18
PubChem CID 8467
ChEBI CHEBI:48408
Formula Weight 166.18
Physical Form Powder
Chemical Name or Material 3-Ethoxy-4-hydroxybenzaldehyde
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.