missing translation for 'onlineSavingsMsg'
Learn More

3-(Di-tert-butylphosphonium)propane sulfonate, 97%, Thermo Scientific Chemicals

Catalog Number p-7032693
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1055888-89-5
C11H25O3PS
268.352
MFCD12546062
JPNPRWMRUCIEMN-UHFFFAOYSA-N
3-di-t-butylphosphonium propane sulfonate, 3-di-tert-butylphosphino propane-1-sulfonic acid, dtbpps, di-t-butyl 3-sulfonatopropyl phosphine, 3-di-tert-butylphosphonium propane sulfonate, 3-di-tert-butylphosphanyl propane-1-sulfonic acid
24949188
3-ditert-butylphosphanylpropane-1-sulfonic acid
CC(C)(C)P(CCCS(=O)(=O)O)C(C)(C)C
This item is not returnable. View return policy
1055888-89-5
C11H25O3PS
268.352
MFCD12546062
JPNPRWMRUCIEMN-UHFFFAOYSA-N
3-di-t-butylphosphonium propane sulfonate, 3-di-tert-butylphosphino propane-1-sulfonic acid, dtbpps, di-t-butyl 3-sulfonatopropyl phosphine, 3-di-tert-butylphosphonium propane sulfonate, 3-di-tert-butylphosphanyl propane-1-sulfonic acid
24949188
3-ditert-butylphosphanylpropane-1-sulfonic acid
CC(C)(C)P(CCCS(=O)(=O)O)C(C)(C)C

3-(Di-tert-butylphosphino)propane-1-sulfonic acid is used in palladium catalyzed cross-coupling reactions. It is also used in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(Di-tert-butylphosphino)propane-1-sulfonic acid is used in palladium catalyzed cross-coupling reactions. It is also used in organic synthesis.

Solubility
Slightly soluble in water.

Notes
Moisture and air sensitive. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Linear Formula [(CH3)3C]2P(CH2)3SO3H
Quantity 250 mg
Sensitivity Air and moisture sensitive
Solubility Information Slightly soluble in water.
Formula Weight 268.35
Percent Purity 97%
Chemical Name or Material 3-(Di-tert-butylphosphonium)propane sulfonate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'