missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorophenylhydrazine hydrochloride, 97%, Thermo Scientific Chemicals

Catalog Number p-7022140
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
2312-23-4
C6H8Cl2N2
179.044
MFCD00012935
CRRIAWUJYMLJOE-UHFFFAOYSA-N
3-chlorophenylhydrazine hydrochloride, 3-chlorophenyl hydrazine hydrochloride, 3-chloro phenyl hydrazine hydrochloride, m-chlorophenylhydrazine hydrochloride, hydrazine, 3-chlorophenyl-, monohydrochloride, 1-3-chlorophenyl hydrazine hydrochloride, m-chlorophenylhydrazine hcl, 3-chlorophenyl hydrazine hydrochloride 1:1, 3-chlorophenyl hydrazinehydrochloride
75331
(3-chlorophenyl)hydrazine;hydrochloride
C1=CC(=CC(=C1)Cl)NN.Cl
This item is not returnable. View return policy
2312-23-4
C6H8Cl2N2
179.044
MFCD00012935
CRRIAWUJYMLJOE-UHFFFAOYSA-N
3-chlorophenylhydrazine hydrochloride, 3-chlorophenyl hydrazine hydrochloride, 3-chloro phenyl hydrazine hydrochloride, m-chlorophenylhydrazine hydrochloride, hydrazine, 3-chlorophenyl-, monohydrochloride, 1-3-chlorophenyl hydrazine hydrochloride, m-chlorophenylhydrazine hcl, 3-chlorophenyl hydrazine hydrochloride 1:1, 3-chlorophenyl hydrazinehydrochloride
75331
(3-chlorophenyl)hydrazine;hydrochloride
C1=CC(=CC(=C1)Cl)NN.Cl

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point ∼242°C (decomposition)
Odor Odorless
Quantity 10 g
Beilstein 3565828
Formula Weight 179.05
Percent Purity 97%
Chemical Name or Material 3-Chlorophenylhydrazine hydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'