Learn More
3-Chlorophenylacetonitrile, 99%, Thermo Scientific Chemicals
Catalog Number 11487607
Click to view available options
Quantity:
5 g
25 g
100 g
1529-41-5
C8H6ClN
151.593
MFCD00001908
GTIKLPYCSAMPNG-UHFFFAOYSA-N
3-chlorobenzyl cyanide, 3-chlorophenyl acetonitrile, 2-3-chlorophenyl acetonitrile, 3-chlorophenylacetonitrile, 3-chlorobenzeneacetonitrile, m-chlorobenzyl cyanide, acetonitrile, m-chlorophenyl, benzeneacetonitrile, 3-chloro, m-chlorophenyl acetonitrile, 2-3-chlorophenyl ethanenitrile
73722
2-(3-chlorophenyl)acetonitrile
C1=CC(=CC(=C1)Cl)CC#N
1529-41-5
C8H6ClN
151.593
MFCD00001908
GTIKLPYCSAMPNG-UHFFFAOYSA-N
3-chlorobenzyl cyanide, 3-chlorophenyl acetonitrile, 2-3-chlorophenyl acetonitrile, 3-chlorophenylacetonitrile, 3-chlorobenzeneacetonitrile, m-chlorobenzyl cyanide, acetonitrile, m-chlorophenyl, benzeneacetonitrile, 3-chloro, m-chlorophenyl acetonitrile, 2-3-chlorophenyl ethanenitrile
73722
2-(3-chlorophenyl)acetonitrile
C1=CC(=CC(=C1)Cl)CC#N
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point 11°C to 13°C
Boiling Point 134°C to 136°C (10 mmHg)
Flash Point >110°C (230°F)
Odor Pungent
Quantity 5 g
UN Number UN3276
Beilstein 2042965
Formula Weight 151.6
Percent Purity 99%
Chemical Name or Material 3-Chlorophenylacetonitrile