Learn More
3-Chloromethyl-5-methylpyridine Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | 3-(Chloromethyl)-5-methylpyridine hydrochloride |
| CAS | 1007089-84-0 |
| Molecular Formula | C7 H8 Cl N . Cl H |
| Quantity | 5 g |
| Synonym | Rupatadine impurity 6 HCl |
| InChI Formula | InChI=1S/C7H8ClN.ClH/c1-6-2-7(3-8)5-9-4-6;/h2,4-5H,3H2,1H3;1H |
| SMILES | Cl.Cc1cncc(CCl)c1 |
| IUPAC Name | 3-(chloromethyl)-5-methylpyridine;hydrochloride |
| Molecular Weight (g/mol) | 178.06 |
| Formula Weight | 177.0112 |