Learn More
3-Chloro-N-[4-cyano-3-(trifluoromethyl)phenyl]-2-hydroxy-2-methylpropanamide, Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
3-Chloro-N-[4-cyano-3-(trifluoromethyl)phenyl]-2-hydroxy-2-methylpropanamide
Specifications
Specifications
| Chemical Name or Material | 3-Chloro-N-[4-cyano-3-(trifluoromethyl)phenyl]-2-hydroxy-2-methylpropanamide |
| CAS | 627484-44-0 |
| Analyte or Component Names | 3-Chloro-N-[4-cyano-3-(trifluoromethyl)phenyl]-2-hydroxy-2-methylpropanamide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C12 H10 Cl F3 N2 O2 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Propanamide, 3-chloro-N-[4-cyano-3-(trifluoromethyl)phenyl]-2-hydroxy-2-methyl-, 3-Chloro-N-[4-cyano-3-(trifluoromethyl)phenyl]-2-hydroxy-2-methylpropanamide |
| InChI Formula | InChI=1S/C12H10ClF3N2O2/c1-11(20,6-13)10(19)18-8-3-2-7(5-17)9(4-8)12(14,15)16/h2-4,20H,6H2,1H3,(H,18,19) |
| SMILES | CC(O)(CCl)C(=O)Nc1ccc(C#N)c(c1)C(F)(F)F |
| IUPAC Name | 3-chloro-N-[4-cyano-3-(trifluoromethyl)phenyl]-2-hydroxy-2-methylpropanamide |
| Show More |