Learn More
3-Chloro-5-(trifluoromethoxy)benzaldehyde, 97%, Thermo Scientific™
Product Code. 11326889
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. 11326889
Brand: Thermo Scientific Alfa Aesar H26788.06
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
3-Chloro-5-(trifluoromethoxy)benzaldehyde is used as pharmaceutical intermediates
Solubility
Soluble in water (3.3 g/L (20°C)).
Notes
Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 433926-48-8
Molecular Formula C8H4ClF3O2
Molecular Weight (g/mol) 224.563
MDL Number MFCD06660305
InChI Key LWQSWEYYGGFLDR-UHFFFAOYSA-N
Synonym 3-chloro-5-trifluoromethoxy benzaldehyde, benzaldehyde, 3-chloro-5-trifluoromethoxy, 5-chloro-3-trifluoromethoxy benzaldehyde
PubChem CID 17750734
IUPAC Name 3-chloro-5-(trifluoromethoxy)benzaldehyde
SMILES C1=C(C=C(C=C1OC(F)(F)F)Cl)C=O
Specifications
Refractive Index 1.4765
Quantity 5 g
Sensitivity Air Sensitive
Solubility Information Soluble in water (3.3g/L (20°C)).
Formula Weight 224.57
Percent Purity 97%
Chemical Name or Material 3-Chloro-5-(trifluoromethoxy)benzaldehyde