missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-(trifluoromethyl)benzylamine, 97%, Thermo Scientific™

Product Code. 15404547
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15404547 1 g 1g
15414547 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15404547 Supplier Thermo Scientific Alfa Aesar Supplier No. H31883.03
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is used as a pharmaceutical intermediate and a primary and secondary intermediate.

Solubility
Insoluble in water.

Notes
Air sensitive. Store in cool dry place. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 361393-93-3
Molecular Formula C8H7ClF3N
Molecular Weight (g/mol) 209.596
MDL Number MFCD04972759
InChI Key GRCRXCBPWHSEOI-UHFFFAOYSA-N
Synonym 3-chloro-4-trifluoromethyl benzylamine, 3-chloro-4-trifluoromethyl benzyl amine, 3-chloro-4-trifluoromethylbenzylamine, 3-chloro-4-trifluoromethyl-benzylamine, 3-chloro-4-trifluoromethyl phenyl methanamine, 1-3-chloro-4-trifluoromethyl phenyl methanamine, benzenemethanamine, 3-chloro-4-trifluoromethyl, pubchem4394, ksc575s7b
PubChem CID 23512744
IUPAC Name [3-chloro-4-(trifluoromethyl)phenyl]methanamine
SMILES C1=CC(=C(C=C1CN)Cl)C(F)(F)F

Specifications

Quantity 1 g
UN Number UN2735
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 209.6
Percent Purity 97%
Chemical Name or Material 3-Chloro-4-(trifluoromethyl)benzylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.