Learn More
3-Chloro-4-methylbenzoic acid, TRC
Product Code. 30357264
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
250mg
500mg
Product Code. Quantity unitSize
30357264 100 mg 100mg
30361395 500 mg 500mg
30356710 250 mg 250mg
3 options
Product Code. 30357264
Brand: Toronto Research Chemicals TRCC587595100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 5162-82-3
Molecular Formula C8H7ClO2
Molecular Weight (g/mol) 170.59
IUPAC Name 3-chloro-4-methylbenzoic acid
SMILES Cc1ccc(cc1Cl)C(=O)O
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C8H7ClO2/c1-5-2-3-6(8(10)11)4-7(5)9/h2-4H,1H3,(H,10,11)
Formula Weight 170.0135
Chemical Name or Material 3-Chloro-4-methylbenzoic acid