Learn More
3-Chloro-4-formylbenzeneboronic acid, 95%, Thermo Scientific™
Product Code. 15423767
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15423767 250 mg 250mg
11322859 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15423767 Supplier Thermo Scientific Alfa Aesar Supplier No. H27725.MD
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
Used in suzuki cross-coupling.
Solubility
Insoluble in water.
Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1072952-53-4
Molecular Formula C7H6BClO3
Molecular Weight (g/mol) 184.382
MDL Number MFCD08274473
InChI Key KDIYVLXLPNHJRZ-UHFFFAOYSA-N
Synonym 3-chloro-4-formylphenyl boronic acid, 3-chloro-4-formylbenzeneboronic acid, acmc-2098uy, ksc496a3d, 3-chloro-4-formylphenyl boronicacid, 3-chloro-4-formyl phenylboronic acid, 4-borono-2-chlorobenzaldehyde, 3-chloro-4-formyl-phenyl boronic acid
PubChem CID 42614540
IUPAC Name (3-chloro-4-formylphenyl)boronic acid
SMILES B(C1=CC(=C(C=C1)C=O)Cl)(O)O
Specifications
Melting Point 210°C to 214°C
Quantity 250 mg
Beilstein EM>3
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 184.39
Percent Purity 95%
Chemical Name or Material 3-Chloro-4-formylbenzeneboronic acid