missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-formylbenzeneboronic acid, 95%, Thermo Scientific™

Product Code. 11322859
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 11322859

Brand: Thermo Scientific Alfa Aesar H27725.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Used in suzuki cross-coupling.

Solubility
Insoluble in water.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1072952-53-4
Molecular Formula C7H6BClO3
Molecular Weight (g/mol) 184.382
MDL Number MFCD08274473
InChI Key KDIYVLXLPNHJRZ-UHFFFAOYSA-N
Synonym 3-chloro-4-formylphenyl boronic acid, 3-chloro-4-formylbenzeneboronic acid, acmc-2098uy, ksc496a3d, 3-chloro-4-formylphenyl boronicacid, 3-chloro-4-formyl phenylboronic acid, 4-borono-2-chlorobenzaldehyde, 3-chloro-4-formyl-phenyl boronic acid
PubChem CID 42614540
IUPAC Name (3-chloro-4-formylphenyl)boronic acid
SMILES B(C1=CC(=C(C=C1)C=O)Cl)(O)O

Specifications

Melting Point 210°C to 214°C
Quantity 1 g
Beilstein EM>3
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 184.39
Percent Purity 95%
Chemical Name or Material 3-Chloro-4-formylbenzeneboronic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.