Learn More
3-Chloro-4-(difluoromethyl)pyridine, 97%, Thermo Scientific™
Product Code. 15427377
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. 15427377
Brand: Thermo Scientific Alfa Aesar H34426.MD
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1374659-44-5
Molecular Formula C6H4ClF2N
Molecular Weight (g/mol) 163.55
MDL Number MFCD22124921
InChI Key UZRBGKNPGFNLDR-UHFFFAOYSA-N
Synonym 3-chloro-4-difluoromethyl pyridine, pyridine, 3-chloro-4-difluoromethyl
PubChem CID 60623021
IUPAC Name 3-chloro-4-(difluoromethyl)pyridine
SMILES FC(F)C1=C(Cl)C=NC=C1
Specifications
Quantity 250 mg
Formula Weight 163.55
Percent Purity 97%
Chemical Name or Material 3-Chloro-4-(difluoromethyl)pyridine