Learn More
3-Chloro-2-methylbenzonitrile, 97%, Thermo Scientific Chemicals
Catalog Number 11327516
Click to view available options
Quantity:
1 g
5 g
54454-12-5
C8H6ClN
151.593
MFCD00045598
FKFZTNLSUJCIMG-UHFFFAOYSA-N
2-methyl-3-chlorobenzonitrile, pubchem23847, 3-chloro-o-tolunitrile, acmc-1aqtx, 6-chloro-2-cyanotoluene, 2-chloro-6-cyanotoluene, 2-methyl-3-chloro-benzonitrile, 3-chloro-2-methyl-benzonitrile, benzonitrile,3-chloro-2-methyl, benzonitrile, 3-chloro-2-methyl
521508
3-chloro-2-methylbenzonitrile
CC1=C(C=CC=C1Cl)C#N
54454-12-5
C8H6ClN
151.593
MFCD00045598
FKFZTNLSUJCIMG-UHFFFAOYSA-N
2-methyl-3-chlorobenzonitrile, pubchem23847, 3-chloro-o-tolunitrile, acmc-1aqtx, 6-chloro-2-cyanotoluene, 2-chloro-6-cyanotoluene, 2-methyl-3-chloro-benzonitrile, 3-chloro-2-methyl-benzonitrile, benzonitrile,3-chloro-2-methyl, benzonitrile, 3-chloro-2-methyl
521508
3-chloro-2-methylbenzonitrile
CC1=C(C=CC=C1Cl)C#N
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point 20°C to 23°C
Boiling Point 120°C (22 mmHg)
Quantity 1 g
Refractive Index 1.553
UN Number UN3439
Beilstein 2613370
Formula Weight 151.6
Percent Purity 97%
Chemical Name or Material 3-Chloro-2-methylbenzonitrile