Learn More
STEMCELL Technologies 3 cc Syringes
Description
- Ideal for accurate dispensing of methylcellulose-based medium (e.g. MethoCult™), when used with Blunt-End Needles (Catalog #28110) purchased from STEMCELL Technologies to ensure compatibility and accuracy
Specifications
Specifications
| For Use With (Application) | Colony Assay |
| Quantity | 100 Syringes/Bag |