missing translation for 'onlineSavingsMsg'
Learn More

3-Carboxy-2-methoxybenzeneboronic acid, 98%, Thermo Scientific Chemicals

Product Code. p-7035604
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15450068

Brand: Thermo Scientific Alfa Aesar H53323.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 913836-10-9
Molecular Formula C8H9BO5
Molecular Weight (g/mol) 195.965
MDL Number MFCD09027237
InChI Key QUWLFFYGKXZCER-UHFFFAOYSA-N
Synonym benzoicacid, 3-borono-2-methoxy, 3-carboxy-2-methoxyphenylboronic acid, 3-dihydroxyboranyl-2-methoxybenzoic acid, 3-carboxy-2-methoxybenzeneboronic acid, acmc-209rau, 2-methoxy-3-dihydroxyboryl benzoic acid
PubChem CID 44119386
IUPAC Name 3-borono-2-methoxybenzoic acid
SMILES B(C1=C(C(=CC=C1)C(=O)O)OC)(O)O

Specifications

Melting Point 152°C to 155°C
Quantity 250 mg
Formula Weight 195.97
Percent Purity 98%
Chemical Name or Material 3-Carboxy-2-methoxybenzeneboronic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.