missing translation for 'onlineSavingsMsg'
Learn More

3-Bromopyruvic acid, MedChemExpress

Product Code. 16451909
Encompass_Preferred
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
Unit Size:
10g
1g
25g
5g
This item is not returnable. View return policy

Product Code. 16451909

Brand: Medchem Express HY19992

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress 3-Bromopyruvate (Bromopyruvic acid) is an analogue of pyruvate and a potent hexokinase (HK)-II inhibitor with high tumor selectivity. 3-Bromopyruvate inhibits cell growth and induces apoptosis through interfering with glycolysis. 3-Bromopyruvate induces autophagy by stimulating ROS formation in breast cancer cells. Antimicrobial activities.

3-Bromopyruvate (Bromopyruvic acid) is an analogue of pyruvate and a potent hexokinase (HK)-II inhibitor with high tumor selectivity. 3-Bromopyruvate inhibits cell growth and induces apoptosis through interfering with glycolysis. 3-Bromopyruvate induces autophagy by stimulating ROS formation in breast cancer cells. Antimicrobial activities.

Specifications

Chemical Name or Material 3-Bromopyruvic acid
Physical Form Solid
CAS 1113-59-3
Color White
Quantity 1 g
Purity Grade Notes Research
Molecular Formula C3H3BrO3
For Use With (Application) Cancer-Kinase/protease
Synonym Bromopyruvic acid Hexokinase II Inhibitor II, 3-BP
Solubility Information H2O : 250 mg/mL (1497.36 mM; Need ultrasonic)
SMILES O=C(O)C(CBr)=O
Molecular Weight (g/mol) 166.96
Formula Weight 166.96
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.