Learn More
3-Bromo-5-(trifluoromethoxy)aniline, 97%, Thermo Scientific™
Product Code. 15465237
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15465237 1 g 1g
15475237 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15465237 Supplier Thermo Scientific Alfa Aesar Supplier No. H32510.03
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 914636-35-4
Molecular Formula C7H5BrF3NO
Molecular Weight (g/mol) 256.022
MDL Number MFCD08458076
InChI Key GJUHRBPRJGNGFH-UHFFFAOYSA-N
Synonym 3-bromo-5-trifluoromethoxy aniline, 3-bromo-5-trifluoromethoxy anilin, 3-bromo-5-trifluoromethoxy benzenamine, 5-bromo-3-trifluoromethoxy phenylamine, 3-amino-5-trifluoromethoxy bromobenzene
PubChem CID 26985421
IUPAC Name 3-bromo-5-(trifluoromethoxy)aniline
SMILES C1=C(C=C(C=C1OC(F)(F)F)Br)N
Specifications
Refractive Index 1.511
Quantity 1 g
UN Number UN2810
Formula Weight 256.02
Percent Purity 97%
Chemical Name or Material 3-Bromo-5-(trifluoromethoxy)aniline