missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-5-nitrobenzoic acid, 99%, Thermo Scientific™

Product Code. 11345425
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11345425 250 mg 250mg
11335425 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11345425 Supplier Thermo Scientific Alfa Aesar Supplier No. L19633.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6307-83-1
Molecular Formula C7H4BrNO4
Molecular Weight (g/mol) 246.016
MDL Number MFCD00100098
InChI Key AXRKIZCFYZBBPX-UHFFFAOYSA-N
Synonym 3-bromo-5-nitrobenzoic acid, 5-bromo-3-nitrobenzoic acid, benzoic acid, 3-bromo-5-nitro, 3-bromo-5-nitro-benzoic acid, 3-bromo-5-nitrobenzoicacid, pubchem4734, acmc-209ncw, ksc357s4t, 3-bromo-5-nitro benzoic acid
PubChem CID 239336
SMILES C1=C(C=C(C=C1[N+](=O)[O-])Br)C(=O)O

Specifications

Melting Point 159°C to 161°C
Quantity 250 mg
Beilstein 2051365
Formula Weight 246.02
Percent Purity 99%
Chemical Name or Material 3-Bromo-5-nitrobenzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.