Learn More
3-Bromo-2-hydroxybenzoic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 3-Bromo-2-hydroxybenzoic Acid |
| CAS | 3883-95-2 |
| Molecular Formula | C31 H62 O3 Si |
| Quantity | 25 g |
| Synonym | 3-Bromosalicylic acid |
| InChI Formula | InChI=1S/C31H62O3Si/c1-9-11-13-15-16-17-18-19-20-22-28(34-35(25(3)4,26(5)6)27(7)8)24-30-29(31(32)33-30)23-21-14-12-10-2/h25-30H,9-24H2,1-8H3/t28-,29+,30?/m1/s1 |
| SMILES | CCCCCCCCCCC[C@H](CC1OC(=O)[C@H]1CCCCCC)O[Si](C(C)C)(C(C)C)C(C)C |
| IUPAC Name | (3S)-3-hexyl-4-[(2R)-2-tri(propan-2-yl)silyloxytridecyl]oxetan-2-one |
| Molecular Weight (g/mol) | 510.908 |
| Formula Weight | 510.447 |