Learn More
3-Bromo-2-fluorophenylacetonitrile, 98%, Thermo Scientific Chemicals
Product Code. 15494687
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. 15494687
Brand: Thermo Scientific Alfa Aesar H32021.MD
Request Bulk or Custom Format
This item requires a price update prior to being ordered.
Request a Price for this item here.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 874285-03-7
Molecular Formula C8H5BrFN
Molecular Weight (g/mol) 214.04
MDL Number MFCD09864662
InChI Key JKRBRQNAVKGEHZ-UHFFFAOYSA-N
Synonym 3-bromo-2-fluorophenylacetonitrile, 3-bromo-2-fluorophenyl acetonitrile, 2-3-bromo-2-fluorophenyl acetonitrile, acmc-20a3z9, benzeneacetonitrile,3-bromo-2-fluoro, 3-bromo-2-fluoro-phenyl-acetonitrile, 2-3-bromo-2-fluorophenyl ethanenitrile
PubChem CID 26985257
IUPAC Name 2-(3-bromo-2-fluorophenyl)acetonitrile
SMILES FC1=C(CC#N)C=CC=C1Br
Specifications
Boiling Point 92°C (0.3 mmHg)
Quantity 250 mg
UN Number UN3276
Formula Weight 214.04
Percent Purity 98%
Chemical Name or Material 3-Bromo-2-fluorophenylacetonitrile