missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-2-cyanopyridine, 98%, Thermo Scientific Chemicals

Product Code. 15463737
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. Quantity unitSize
15463737 250 mg 250mg
11496580 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 15463737

Brand: Thermo Scientific Alfa Aesar H27632.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55758-02-6
Molecular Formula C6H3BrN2
Molecular Weight (g/mol) 183.01
MDL Number MFCD02683288
InChI Key HCOPIUVJCIZALB-UHFFFAOYSA-N
Synonym 3-bromo-2-cyanopyridine, 3-bromopicolinonitrile, 2-cyano-3-bromopyridine, 3-bromo-2-pyridinecarbonitrile, 3-bromo-pyridine-2-carbonitrile, 2-pyridinecarbonitrile, 3-bromo, pubchem2263, 3-bromopicolino-nitrile, bromo-2-cyano-pyridine, 3-bromo-2-cyano-pyridine
PubChem CID 817694
IUPAC Name 3-bromopyridine-2-carbonitrile
SMILES BrC1=CC=CN=C1C#N

Specifications

Melting Point 96°C to 99°C
Boiling Point 120°C (4 mmHg)
Quantity 250 mg
Formula Weight 183.01
Percent Purity 98%
Chemical Name or Material 3-Bromo-2-cyanopyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.