missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-1-benzofuran, 97%, Thermo Scientific™

Product Code. p-4510324
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10710374 250 mg Amber Glass Bottle 250mg
10255792 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10710374 Supplier Thermo Scientific Maybridge Supplier No. CC27510CB

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 59214-70-9
Molecular Formula C8H5BrO
Molecular Weight (g/mol) 197.031
InChI Key ICJNAOJPUTYWNV-UHFFFAOYSA-N
Synonym 3-bromobenzofuran, 3-bromobenzo b furan, benzofuran, 3-bromo, 3-bromo-1-benzo b furan, 3-bromobenzofurane, 3-bromo-benzo b furan, 3-bromanyl-1-benzofuran, ksc269c4b
PubChem CID 640589
IUPAC Name 3-bromo-1-benzofuran
SMILES C1=CC=C2C(=C1)C(=CO2)Br

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 197.03
Percent Purity 97%
Chemical Name or Material 3-Bromo-1-benzofuran
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.