Learn More
3-(Boc-amino)-2-thiophenecarboxylic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 3-(Boc-amino)-2-thiophenecarboxylic Acid |
| CAS | 101537-64-8 |
| Molecular Formula | C10H13NO4S |
| Quantity | 500 mg |
| InChI Formula | InChI=1S/C10H13NO4S/c1-10(2,3)15-9(14)11-6-4-5-16-7(6)8(12)13/h4-5H,1-3H3,(H,11,14)(H,12,13) |
| SMILES | CC(C)(C)OC(=O)Nc1ccsc1C(=O)O |
| IUPAC Name | 3-[(2-methylpropan-2-yl)oxycarbonylamino]thiophene-2-carboxylic acid |
| Molecular Weight (g/mol) | 243.28 |
| Formula Weight | 243.0565 |