Learn More
3'-Azido-3'-deoxy-beta-L-uridine, MedChemExpress
Product Code. 16444508
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Product Code. Quantity unitSize
16444508 10 mg 10mg
16460817 50 mg 50mg
2 options
Product Code. 16444508
Brand: Medchem Express HY111642
MedChemExpress 3'-Azido-3'-deoxy-beta-L-uridine (Compound 25) is a nucleoside derivative.
3'-Azido-3'-deoxy-beta-L-uridine (Compound 25) is a nucleoside derivative.
Specifications
Chemical Name or Material 3'-Azido-3'-deoxy-beta-L-uridine
CAS 2095417-28-8
Color White
Purity Grade Notes Research
Molecular Formula C9H11N5O5
Quantity 10 mg
Solubility Information DMSO : 125 mg/mL (464.32 mM; Need ultrasonic)
SMILES O=C(N([C@H]1O[C@@H](CO)[C@H](N=[N+]=[N-])[C@@H]1O)C=C2)NC2=O
Molecular Weight (g/mol) 269.21
Formula Weight 269.21
Percent Purity 99.7%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation