Learn More
3-Aminopyridine, TRC
Product Code. 30011483
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
Unit Size:
100g
10g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30011483 10 g 10g
30009926 100 g 100g
30010298 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30011483 Supplier Toronto Research Chemicals Supplier No. TRCA62901510G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 462-08-8
Molecular Formula C5 H6 N2
Molecular Weight (g/mol) 94.11
Synonym Pyridine, 3-amino- (8CI), 3-Aminopyridine, 3-Pyridinylamine, 3-Pyridylamine, NSC 15040, m-Aminopyridine, β-Aminopyridine
IUPAC Name pyridin-3-amine
SMILES Nc1cccnc1
Specifications
Quantity 10 g
InChI Formula InChI=1S/C5H6N2/c6-5-2-1-3-7-4-5/h1-4H,6H2
Formula Weight 94.0531
Chemical Name or Material 3-Aminopyridine