Learn More
3-Aminobiphenyl-d9, TRC
Specifications
Specifications
| Chemical Name or Material | 3-Aminobiphenyl-d9 |
| CAS | 1020718-93-7 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C12 2H9 H2 N |
| Quantity | 2.5 mg |
| Synonym | [1,1'-Biphenyl-2,2',3',4,4',5,5',6,6'-d9]-3-amine, 3-Biphenylamine (ring-D9), (1,1'-Biphenyl-3-yl)amine (ring-D9), 3-Amino-2,2',3',4,4',5,5',6,6'-nonadeuterio-1,1'-biphenyl, 3-Aminobiphenyl (ring-D9), 3-Phenylaniline (ring-D9), m-Aminobiphenyl (ring-D9), m-Phenylaniline (ring-D9) |
| InChI Formula | InChI=1S/C12H11N/c13-12-8-4-7-11(9-12)10-5-2-1-3-6-10/h1-9H,13H2/i1D,2D,3D,4D,5D,6D,7D,8D,9D |
| SMILES | [2H]c1c([2H])c([2H])c(c([2H])c1[2H])c2c([2H])c([2H])c([2H])c(N)c2[2H] |
| IUPAC Name | 2,3,4,6-tetradeuterio-5-(2,3,4,5,6-pentadeuteriophenyl)aniline |
| Molecular Weight (g/mol) | 178.28 |
| Show More |