Learn More
3-Amino-5-nitrobenzeneboronic acid hydrochloride, 96%, Thermo Scientific™
Product Code. 15400016
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15400016 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15400016 Supplier Thermo Scientific Alfa Aesar Supplier No. H52674.MD
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Specifications
CAS 389621-79-8
Melting Point 136°C to 141°C
Molecular Formula C6H8BClN2O4
MDL Number MFCD03411933
Quantity 250 mg
Synonym 3-amino-5-nitrophenyl boronic acid hydrochloride, 3-amino-5-nitrophenylboronic acid, hcl, 3-amino-5-nitrophenylboronic acid hydrochloride, 3-amino-5-nitrophenyl boronic acid hcl salt, 3-amino-5-nitrophenyl boronic acid hydrochloride salt, 3-amino-5-nitrophenyl boronic acid hcl, 3-amino-5-nitrobenzeneboronic acid hydrochloride, 3-amino-5-nitrophenyboronic acid hcl salt, 3-azanyl-5-nitro-phenyl boronic acid hydrochloride, 3-amino-5-nitrophenyl-boronic acid monohydrochloride
InChI Key QRMMGONMBRVVKK-UHFFFAOYSA-N
SMILES B(C1=CC(=CC(=C1)[N+](=O)[O-])N)(O)O.Cl
IUPAC Name (3-amino-5-nitrophenyl)boronic acid;hydrochloride
Molecular Weight (g/mol) 218.4
PubChem CID 16427089
Formula Weight 218.4
Percent Purity 96%
Chemical Name or Material 3-Amino-5-nitrobenzeneboronic acid hydrochloride