Learn More
3-Amino-5-fluoropyridine, 98%, Thermo Scientific Chemicals
Product Code. 15487027
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
15487027 5 g 5g
15477027 1 g 1g
2 options
Product Code. 15487027
Brand: Thermo Scientific Alfa Aesar H34111.06
Request Bulk or Custom Format
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 210169-05-4
Molecular Formula C5H5FN2
Molecular Weight (g/mol) 112.107
MDL Number MFCD00234073
InChI Key ZRORIJXOWXYPMO-UHFFFAOYSA-N
Synonym 3-amino-5-fluoropyridine, 5-amino-3-fluoropyridine, 3-pyridinamine, 5-fluoro, 5-fluoro-3-pyridylamine, 5-fluoro-3-pyridinamine, 3-fluoro-5-aminopyridine, 5-fluoropyridin-3-ylamine, 3-amine-5-fluoro-pyridin, 3-amino-5-fluoro-pyridin, 3-amine-5-fluoro-pyridine
PubChem CID 574450
IUPAC Name 5-fluoropyridin-3-amine
SMILES C1=C(C=NC=C1F)N
Specifications
Melting Point 85°C to 89°C
Quantity 5 g
Formula Weight 112.11
Percent Purity 98%
Chemical Name or Material 3-Amino-5-fluoropyridine