Learn More
3-Amino-4,4,4-trifluorobutyric acid, 97%
Product Code. 11338695
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11338695 250 mg 250mg
11328695 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11338695 Supplier Thermo Scientific Alfa Aesar Supplier No. L12239.MD
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
CAS: 584-20-3 | C4H6F3NO2 | 157.09 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 584-20-3
Molecular Formula C4H6F3NO2
Molecular Weight (g/mol) 157.09
MDL Number MFCD00041413
InChI Key XRXQOEWWPPJVII-REOHCLBHSA-N
PubChem CID 342970
IUPAC Name 3-amino-4,4,4-trifluorobutanoic acid
SMILES [NH3+][C@@H](CC([O-])=O)C(F)(F)F
Specifications
Melting Point ∽184°C (decomposition)
Beilstein 1764276
Synonym 3-amino-4,4,4-trifluorobutyric acid, butanoic acid, 3-amino-4,4,4-trifluoro, butanoic acid, 3-amino-4,4,4-trifluoro-, 3r, acmc-1ajx7, acmc-20n6c3, 3-trifluoromethyl-beta-alanine, butanoicacid,3-amino-4,4,4-trifluoro, butanoic acid,3-amino-4,4,4-trifluoro, r-3-amino-4,4,4-trifluorobutyric acid
Quantity 250 mg
Formula Weight 157.09
Percent Purity 97%
Chemical Name or Material 3-Amino-4,4,4-trifluorobutyric acid