Learn More
3-Amino-3-phenylpropionic acid, 99%
Product Code. 11309775
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. 11309775
Brand: Thermo Scientific Alfa Aesar L19801.03
Request Bulk or Custom Format
CAS: 614-19-7 | C9H11NO2 | 165.19 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 614-19-7
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.19
MDL Number MFCD00008064,MFCD01076238
InChI Key UJOYFRCOTPUKAK-UHFFFAOYNA-N
Synonym 3-amino-3-phenylpropionic acid, dl-3-amino-3-phenylpropionic acid, dl-beta-phenylalanine, beta-phenyl-beta-alanine, 3-phenyl-dl-beta-alanine, h-dl-beta-phe-oh, benzenepropanoic acid, beta-amino, 3-amino-3-phenyl-propionic acid, 3a3ppa, beta-aminohydrocinnamic acid
PubChem CID 69189
ChEBI CHEBI:68528
IUPAC Name 3-amino-3-phenylpropanoic acid
SMILES NC(CC(O)=O)C1=CC=CC=C1
Specifications
Melting Point ∽220°C (decomposition)
Beilstein 2803286
Quantity 1 g
Formula Weight 165.19
Percent Purity 99%
Chemical Name or Material 3-Amino-3-phenylpropionic acid