Learn More
3-Amino-2,5-dichlorobenzoic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | Chloramben |
| CAS | 133-90-4 |
| Molecular Formula | C7 H5 Cl2 N O2 |
| Quantity | 2.5 g |
| Synonym | 3-Amino-2,5-dichlorobenzoic acid, 2,5-Dichloro-3-aminobenzoic acid, Ambiben, Amiben, Amibin, Amoben, Chloramben, Vegiben |
| InChI Formula | InChI=1S/C7H5Cl2NO2/c8-3-1-4(7(11)12)6(9)5(10)2-3/h1-2H,10H2,(H,11,12) |
| SMILES | Nc1cc(Cl)cc(C(=O)O)c1Cl |
| IUPAC Name | 3-amino-2,5-dichlorobenzoic acid |
| Molecular Weight (g/mol) | 206.03 |
| Formula Weight | 204.9697 |