missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ (+/-)-3-Amino-1-(benzyloxycarbonyl)pyrrolidine

Product Code. p-7030415
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15403827 250 mg 250mg
11472070 1 g 1g
11482070 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15403827 Supplier Alfa Aesar™ Supplier No. H28525.MD
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 185057-50-5
Molecular Formula C12H16N2O2
Molecular Weight (g/mol) 220.272
MDL Number MFCD03001673
InChI Key FPXJNSKAXZNWMQ-UHFFFAOYSA-N
Synonym 3-amino-1-n-cbz-pyrrolidine, 1-cbz-3-aminopyrrolidine, 1-cbz-3-amino pyrrolidine, 1-benzyloxycarbonyl-3-aminopyrrolidine, 3-amino-pyrrolidine-1-carboxylic acid benzyl ester, 1-n-cbz-3-aminopyrrolidine, 3-amino-1-cbz-pyrrolidine, phenylmethyl 3-aminopyrrolidinecarboxylate, 1-pyrrolidinecarboxylic acid, 3-amino-, phenylmethyl ester, n-cbz-3-amino-pyrrolidine
PubChem CID 2756614
IUPAC Name benzyl 3-aminopyrrolidine-1-carboxylate
SMILES C1CN(CC1N)C(=O)OCC2=CC=CC=C2

Specifications

Refractive Index 1.5485
Quantity 250 mg
Sensitivity Air Sensitive
Formula Weight 220.27
Chemical Name or Material (+/-)-3-Amino-1-(benzyloxycarbonyl)pyrrolidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.