Learn More
3-Acetylpyridine N-Oxide, TRC
Specifications
Specifications
| Chemical Name or Material | 3-Acetylpyridine N-Oxide |
| CAS | 14188-94-4 |
| Molecular Formula | C7H7NO2 |
| Quantity | 10 mg |
| Synonym | 1-(1-Oxido-3-pyridinyl)-ethanone, 1-(3-Pyridinyl)-ethanone N-Oxide, Methyl 3-Pyridyl Ketone 1-Oxide, Methyl 3-Pyridyl Ketone N-Oxide, 1-(1-Oxido-3-pyridinyl)ethanone, 1-(1-Oxopyridin-3-yl)ethanone, 3-Acetylpyridine 1-Oxide |
| InChI Formula | InChI=1S/C7H7NO2/c1-6(9)7-3-2-4-8(10)5-7/h2-5H,1H3 |
| SMILES | CC(=O)c1ccc[n+]([O-])c1 |
| IUPAC Name | 1-(1-oxidopyridin-1-ium-3-yl)ethanone |
| Molecular Weight (g/mol) | 137.14 |
| Formula Weight | 137.0477 |