missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies 3% Acetic Acid with Methylene Blue

Product Code. 17190002
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
Product Code. Quantity unitSize
17190002 100 mL 100mL
1 options
This item is not returnable. View return policy

Product Code. 17190002

Brand: STEMCELL Technologies 07060

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Reagent for manual counting of nucleated mammalian cells

  • 3% Acetic Acid with Methylene Blue is recommended for manual counting of nucleated mammalian cells
  • It will lyse red blood cell and white blood cell membranes
  • The remaining white blood cell nuclei will stain lightly with methylene blue

Specifications

Cell Type Other
For Use With (Application) Cell Culture
Quantity 100 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.