Learn More
3,7-Dimethyl-1-octanol, 95%
Alle ansehen Thermo Scientific Produkte
missing translation for 'orderingAttributeHoverText'
Quantity:
50 mL
250 mL
1 L
Packaging:
Glass bottle
missing translation for 'unitSize'
1 L
250mL
50mL
Beschreibung
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spezifikation
Spezifikation
| CAS Min % | 94.0 |
| CAS Max % | 100.0 |
| Color | Colorless |
| Density | 0.8280g/mL |
| Boiling Point | 118°C (15.0 mmHg) |
| Flash Point | 97°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 94% min. (GC) |
| Packaging | Glass bottle |
| Linear Formula | (CH3)2CH(CH2)3CH(CH3)(CH2)2OH |
| Mehr anzeigen |
Sicherheit und Handhabung
Produktidentifikator
- 3,7-Dimethyl-1-octanol
- Warning
- LONG-TERM AQUATIC HAZARD Chronic 2
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- H319-Causes serious eye irritation.
- H411-Toxic to aquatic life with long lasting effects.
- H315-Causes skin irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.