Learn More
3,7,11,15-Tetramethylhexadec-1-en-3-ol, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 505-32-8 |
| Molecular Formula | C20H40O |
| MDL Number | MFCD00048380 |
| Quantity | 100 g |
| SMILES | CC(CCCC(CCCC(C)C)C)CCCC(C)(C=C)O |
| Molecular Weight (g/mol) | 296.53 |
| Percent Purity | 98% |
| Chemical Name or Material | 3,7,11,15-Tetramethylhexadec-1-en-3-ol |