missing translation for 'onlineSavingsMsg'
Learn More

3,6-Dichloro-2-fluorobenzoyl chloride, 97%, Thermo Scientific™

Catalog Number 15485647
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
916420-63-8
C7H2Cl3FO
227.44
MFCD09025383
HJKPBFBHBLRYPL-UHFFFAOYSA-N
46737549
3,6-dichloro-2-fluorobenzoyl chloride
C1=CC(=C(C(=C1Cl)C(=O)Cl)F)Cl
This item is not returnable. View return policy
916420-63-8
C7H2Cl3FO
227.44
MFCD09025383
HJKPBFBHBLRYPL-UHFFFAOYSA-N
46737549
3,6-dichloro-2-fluorobenzoyl chloride
C1=CC(=C(C(=C1Cl)C(=O)Cl)F)Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Refractive Index 1.5467
Quantity 1 g
UN Number UN3265
Sensitivity Moisture sensitive
Formula Weight 227.45
Percent Purity 97%
Chemical Name or Material 3,6-Dichloro-2-fluorobenzoyl chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'