missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-Dimethylanisole, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Descrizione
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifica
CAS Min % | 98.5 |
Color | Colorless to Yellow |
CAS Max % | 100.0 |
Density | 0.9600g/mL |
Boiling Point | 193°C |
Quantity | 25 g |
Infrared Spectrum | Authentic |
Packaging | Glass bottle |
Physical Form | Liquid |
Refractive Index | 1.5117 to 1.5137 |
Vedi altri risultati |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'