Learn More
3,5-Dihydroxytoluene, Sigma-Aldrich™
Product Code. 15861271
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15861271 100 g 100g
15841271 5 g 5g
15851271 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15861271 Supplier Sigma Aldrich Supplier No. 8209330100
 Restricted Item
Estimated Shipment: 18-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 504-15-4
Grade Research
Molecular Weight (g/mol) 124.14
Specifications
Melting Point 109°C
Boiling Point 147°C/7 hPa
MDL Number MFCD00002291
Quantity 100 g
Synonym 3,5-Dihydroxytoluene, 5-Methylresorcinol, Orcinol, 3,5-Dihydroxytoluene