Learn More
3,5-Dichloroaniline, Sigma-Aldrich™
Specifications
Specifications
| CAS | 626-43-7 |
| Melting Point | 46°C to 53°C |
| Density | 1.58 g/cm3 at 20°C |
| Boiling Point | 259°C to 260°C/1013 hPa |
| MDL Number | MFCD00007774 |
| Quantity | 100 g |
| Synonym | 3,5-Dichloroaniline, 3,5-Dichloroaniline |
| Molecular Weight (g/mol) | 162.02 |
| Grade | Research |