Learn More
3,5-Dichloro-2,6-dimethoxybenzoic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 3,5-Dichloro-2,6-dimethoxybenzoic Acid |
| CAS | 73219-91-7 |
| Molecular Formula | C9H8Cl2O4 |
| Quantity | 10 g |
| Synonym | 3,5-Dichloro-2,6-dimethoxybenzoic Acid |
| InChI Formula | InChI=1S/C9H8Cl2O4/c1-14-7-4(10)3-5(11)8(15-2)6(7)9(12)13/h3H,1-2H3,(H,12,13) |
| SMILES | COc1c(Cl)cc(Cl)c(OC)c1C(=O)O |
| IUPAC Name | 3,5-dichloro-2,6-dimethoxybenzoic acid |
| Molecular Weight (g/mol) | 251.06 |
| Formula Weight | 249.98 |