missing translation for 'onlineSavingsMsg'
Learn More

3,5-Di-tert-butyl-4-hydroxyphenylpropionic acid, 98%

Product Code. p-7025085
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11434347

Brand: Thermo Scientific Alfa Aesar B20412.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 20170-32-5 | C17H26O3 | 278.392 g/mol

It is employed as an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as an active pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20170-32-5
MDL Number MFCD00017519
Synonym 3-3,5-di-tert-butyl-4-hydroxyphenyl propionic acid, 3-3,5-di-tert-butyl-4-hydroxyphenyl propanoic acid, fenozan, 3,5-di-tert-butyl-4-hydroxyphenylpropionic acid, benzenepropanoic acid, 3,5-bis 1,1-dimethylethyl-4-hydroxy, unii-x6g09g700f, 3,5-di-tert-butyl-4-hydroxyhydocinnamic acid, 3,5-bis 1,1-dimethylethyl-4-hydroxybenzenepropanoic acid, 3-3,5-ditertbutyl-4-hydroxyphenyl propionic acid, 3-3,5-bis tert-butyl-4-hydroxyphenyl propanoic acid
PubChem CID 88389
IUPAC Name 3-(3,5-ditert-butyl-4-hydroxyphenyl)propanoic acid

Specifications

Melting Point 172°C to 175°C
Molecular Formula C17H26O3
Quantity 5 g
Solubility Information Slightly soluble in water.
InChI Key WPMYUUITDBHVQZ-UHFFFAOYSA-N
SMILES CC(C)(C)C1=CC(=CC(=C1O)C(C)(C)C)CCC(=O)O
Molecular Weight (g/mol) 278.392
Formula Weight 278.4
Percent Purity 98%
Chemical Name or Material 3,5-Di-tert-butyl-4-hydroxyphenylpropionic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.