missing translation for 'onlineSavingsMsg'
Learn More

3,5-Di-tert-butyl-4-hydroxybenzyl alcohol, 97%

Product Code. 11409120
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11409120 100 g 100g
11489110 5 g 5g
11499110 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11409120 Supplier Thermo Scientific Alfa Aesar Supplier No. A14709.22
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 88-26-6 | C15H24O2 | 236.355 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88-26-6
Molecular Formula C15H24O2
Molecular Weight (g/mol) 236.355
MDL Number MFCD00017254
InChI Key HNURKXXMYARGAY-UHFFFAOYSA-N
Synonym 3,5-di-tert-butyl-4-hydroxybenzyl alcohol, 2,6-di-tert-butyl-4-hydroxymethyl phenol, 2,6-di-tert-butyl-4-hydroxymethylphenol, bht alcohol, antioxidant 754, ionox 100, ionox 100 antioxidant, 3,5-di-t-butyl-4-hydroxybenzyl alcohol, butylated hydroxymethylphenol, 2,6-di-t-butyl-4-hydroxymethylphenol
PubChem CID 6929
IUPAC Name 2,6-ditert-butyl-4-(hydroxymethyl)phenol
SMILES CC(C)(C)C1=CC(=CC(=C1O)C(C)(C)C)CO

Specifications

Melting Point 138°C to 141°C
Boiling Point 214°C (40 mmHg)
Quantity 100 g
Beilstein 2052291
Formula Weight 236.36
Percent Purity 97%
Chemical Name or Material 3,5-Di-tert-butyl-4-hydroxybenzyl alcohol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.