missing translation for 'onlineSavingsMsg'
Learn More

3-[4-(Methylsulfonyl)phenyl]propionic acid, 96%, Thermo Scientific™

Product Code. 15486617
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
This item is not returnable. View return policy

Product Code. 15486617

Brand: Thermo Scientific Alfa Aesar H33764.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 387350-46-1
Molecular Formula C10H12O4S
Molecular Weight (g/mol) 228.26
MDL Number MFCD01319236
InChI Key ZZZBVBATZOAPQB-UHFFFAOYSA-N
Synonym 3-4-methylsulfonyl phenyl propanoic acid, 3-4-methanesulfonylphenyl propanoic acid, 3-4-methylsulphonylphenyl propionic acid, benzenepropanoic acid, 4-methylsulfonyl, 4-methylsulfonyl benzenepropanoic acid, 3-4-methylsulfonylphenyl propanoic acid, 3-4-methylsulfonyl benzene-1-propanoic acid, 3-4-methylsulfonyl phenyl propionic acid, 3-4-methylsulphonyl phenyl propanoic acid
PubChem CID 2736960
IUPAC Name 3-(4-methylsulfonylphenyl)propanoic acid
SMILES CS(=O)(=O)C1=CC=C(CCC(O)=O)C=C1

Specifications

Melting Point 173°C to 177°C
Quantity 1 g
Formula Weight 228.26
Percent Purity 96%
Chemical Name or Material 3-[4-(Methylsulfonyl)phenyl]propionic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.