Learn More
3-(4-Methyl-1H-imidazol-1-yl)-5-trifluoromethylaniline, Mikromol™
Description
3-(4-Methyl-1H-imidazol-1-yl)-5-trifluoromethylaniline
Specifications
Specifications
| Chemical Name or Material | 3-(4-Methyl-1H-imidazol-1-yl)-5-trifluoromethylaniline |
| CAS | 641571-11-1 |
| Analyte or Component Names | 3-(4-Methyl-1H-imidazol-1-yl)-5-trifluoromethylaniline |
| Shipping Condition | Room Temperature |
| Molecular Formula | C11 H10 F3 N3 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Benzenamine, 3-(4-methyl-1H-imidazol-1-yl)-5-(trifluoromethyl)-, 3-(4-Methyl-1H-imidazol-1-yl)-5-(trifluoromethyl)benzenamine, 1-(3-Amino-5-trifluoromethylphenyl)-4-methylimidazole, 3-(4-Methyl-1H-imidazol-1-yl)-5-trifluoromethylaniline, 3-(4-Methylimidazol-1-yl)-5-trifluoromethylphenylamine, 5-(4-Methyl-1H-imidazol-1-yl)-3-(trifluoromethyl)benzenamine, 5-(4-Methyl-1H-imidazol-1-yl)-3-(trifluoromethyl)benzeneamine, Nilotinib Hydrochloride Monohydrate Impurity A, Nilotinib Impurity A |
| InChI Formula | InChI=1S/C11H10F3N3/c1-7-5-17(6-16-7)10-3-8(11(12,13)14)2-9(15)4-10/h2-6H,15H2,1H3 |
| SMILES | Cc1cn(cn1)c2cc(N)cc(c2)C(F)(F)F |
| IUPAC Name | 3-(4-methylimidazol-1-yl)-5-(trifluoromethyl)aniline |
| Show More |