missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-(4-Methoxyphenyl)-2-phenylpropionic acid

Product Code. 11449320
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 11449320

Brand: Alfa Aesar™ H50213.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 4314-68-5
Molecular Formula C16H16O3
Molecular Weight (g/mol) 256.301
MDL Number MFCD00046532
InChI Key GIROORGDVPRQFZ-UHFFFAOYSA-N
Synonym 3-4-methoxyphenyl-2-phenylpropanoic acid, 3-4-methoxy-phenyl-2-phenyl-propionic acid, 3-4-methoxyphenyl-2-phenylpropionic acid, chembl83949, acmc-20anjp, benzenepropanoic acid,4-methoxy-a-phenyl, 4-methoxy-alpha-phenylbenzenepropanoic acid, 3-4-methoxyphenyl-2-phenyl-propanoic acid, 3-p-methoxyphenyl-2-phenylpropionic acid
PubChem CID 97739
IUPAC Name 3-(4-methoxyphenyl)-2-phenylpropanoic acid
SMILES COC1=CC=C(C=C1)CC(C2=CC=CC=C2)C(=O)O

Specifications

Quantity 1 g
Formula Weight 256.3
Chemical Name or Material 3-(4-Methoxyphenyl)-2-phenylpropionic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.