Learn More
3-(4-Hydroxyphenyl)-1-propanol, TRC
Product Code. 30403778
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30403778 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30403778 Supplier Toronto Research Chemicals Supplier No. TRCH9528301G
 Restricted Item
Estimated Shipment: 12-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 3-(4-Hydroxyphenyl)-1-propanol
CAS 10210-17-0
Molecular Formula C9H12O2
Quantity 1 g
Synonym 1-(4-Hydroxyphenyl)-3-propanol, 3-(4-Hydroxyphenyl)-1-propanol, 3-(4-Hydroxyphenyl)propanol, 3-(p-Hydroxyphenyl)-1-propanol, 3-(p-Hydroxyphenyl)propyl alcohol, 4-(3-Hydroxypropyl)phenol, 4-Hydroxybenzenepropanol, Dihydro-p-coumaryl Alcohol
InChI Formula InChI=1S/C9H12O2/c10-7-1-2-8-3-5-9(11)6-4-8/h3-6,10-11H,1-2,7H2
SMILES OCCCc1ccc(O)cc1
IUPAC Name 4-(3-hydroxypropyl)phenol
Molecular Weight (g/mol) 152.19
Formula Weight 152.0837